Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTCGGTTGTGTTTTGTACCAT[C/T]TTCTTGAGGAAAACGTTGTGTAATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601865 | |||||||||||||||||||||||
Literature Links: |
PLOD2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLOD2 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000935.2 | 2901 | UTR 3 | NP_000926.2 | |||
NM_182943.2 | 2901 | UTR 3 | NP_891988.1 | |||
XM_005247535.3 | 2901 | UTR 3 | XP_005247592.1 | |||
XM_017006625.1 | 2901 | UTR 3 | XP_016862114.1 |