Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAATCTGAAGGGATGATGGATG[A/C]ATCAAAAGGTGAGTGGGTGAAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600585 | ||||||||||||||||||||
Literature Links: |
TGM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TGM4 - transglutaminase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003241.3 | 84 | Missense Mutation | GAA,GCA | E,A 4 | NP_003232.2 | |
XM_011534042.1 | 84 | Missense Mutation | GAA,GCA | E,A 4 | XP_011532344.1 |
TMEM42 - transmembrane protein 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |