Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGATTCCGGCTCACGGCTATTC[A/G]GCCGTGGGTCCCTGGCAGAAGTTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 125290 MIM: 605474 MIM: 607433 | |||||||||||||||||||||||
Literature Links: |
ALAS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ALAS1 - 5'-aminolevulinate synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLR9 - toll like receptor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017442.3 | 3727 | Silent Mutation | GCC,GCT | A,A 1031 | NP_059138.1 |
TWF2 - twinfilin actin binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |