Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCGCGTGGCTGTCCTGTTCAC[C/T]CGTTCTGAGGATGCCCGGGAGCTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604099 | |||||||||||||||||||||||
Literature Links: |
GRM2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GRM2 - glutamate metabotropic receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000839.3 | 1527 | Silent Mutation | ACC,ACT | T,T 270 | NP_000830.2 | |
NM_001130063.1 | 1527 | Intron | NP_001123535.1 | |||
XM_011533635.1 | 1527 | Silent Mutation | ACC,ACT | T,T 270 | XP_011531937.1 | |
XM_011533636.1 | 1527 | Intron | XP_011531938.1 | |||
XM_011533637.1 | 1527 | Silent Mutation | ACC,ACT | T,T 76 | XP_011531939.1 | |
XM_011533638.2 | 1527 | Silent Mutation | ACC,ACT | T,T 76 | XP_011531940.1 | |
XM_011533639.2 | 1527 | Silent Mutation | ACC,ACT | T,T 76 | XP_011531941.1 | |
XM_011533640.2 | 1527 | Silent Mutation | ACC,ACT | T,T 76 | XP_011531942.1 | |
XM_011533641.2 | 1527 | Silent Mutation | ACC,ACT | T,T 76 | XP_011531943.1 | |
XM_011533642.1 | 1527 | Silent Mutation | ACC,ACT | T,T 270 | XP_011531944.1 | |
XM_011533643.1 | 1527 | Intron | XP_011531945.1 | |||
XM_011533644.1 | 1527 | Intron | XP_011531946.1 | |||
XM_011533645.1 | 1527 | Intron | XP_011531947.1 | |||
XM_011533646.2 | 1527 | Intron | XP_011531948.1 | |||
XM_011533647.1 | 1527 | Intron | XP_011531949.1 | |||
XM_017006271.1 | 1527 | Silent Mutation | ACC,ACT | T,T 270 | XP_016861760.1 |
TEX264 - testis expressed 264 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |