Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAGATTATCAGGAGTGTGAACT[C/T]GCAAGCCATCAGTTAACTGATGGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602719 | |||||||||||||||||||||||
Literature Links: |
TRA2B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRA2B - transformer 2 beta homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243879.1 | 3057 | UTR 3 | NP_001230808.1 | |||
NM_004593.2 | 3057 | UTR 3 | NP_004584.1 | |||
XM_005247703.1 | 3057 | Intron | XP_005247760.1 | |||
XM_011513072.2 | 3057 | Intron | XP_011511374.1 | |||
XM_017007033.1 | 3057 | Intron | XP_016862522.1 |