Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTACTCTCCCTGTCAGTTCCAG[C/T]CCTACCTGACGCCCCACAGACCTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614995 | |||||||||||||||||||||||
Literature Links: |
IL17RE PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IL17RE - interleukin 17 receptor E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193380.1 | Intron | NP_001180309.1 | ||||
NM_153480.1 | Intron | NP_705613.1 | ||||
NM_153481.1 | Intron | NP_705614.1 | ||||
NM_153483.2 | Intron | NP_705616.2 | ||||
XM_006712975.1 | Intron | XP_006713038.1 | ||||
XM_006712976.1 | Intron | XP_006713039.1 | ||||
XM_011533361.2 | Intron | XP_011531663.1 | ||||
XM_011533362.2 | Intron | XP_011531664.1 | ||||
XM_011533363.2 | Intron | XP_011531665.1 | ||||
XM_011533364.2 | Intron | XP_011531666.1 |