Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATTCTCAGCAGCCACTCTTTGG[A/T]CTCGTTCTTCTTCCAGCAGCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602500 MIM: 601306 | ||||||||||||||||||||
Literature Links: |
GOLGB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOLGB1 - golgin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256486.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3172 | NP_001243415.1 | |
NM_001256487.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3128 | NP_001243416.1 | |
NM_001256488.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3087 | NP_001243417.1 | |
NM_004487.4 | 9990 | Missense Mutation | GAC,GTC | D,V 3162 | NP_004478.3 | |
XM_005247371.4 | 9990 | Missense Mutation | GAC,GTC | D,V 3172 | XP_005247428.1 | |
XM_005247372.4 | 9990 | Missense Mutation | GAC,GTC | D,V 3133 | XP_005247429.1 | |
XM_005247373.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3131 | XP_005247430.1 | |
XM_006713587.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3167 | XP_006713650.1 | |
XM_006713588.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3167 | XP_006713651.1 | |
XM_006713589.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3166 | XP_006713652.1 | |
XM_006713590.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3126 | XP_006713653.1 | |
XM_006713591.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3092 | XP_006713654.1 | |
XM_011512699.2 | 9990 | Missense Mutation | GAC,GTC | D,V 3172 | XP_011511001.1 | |
XM_017006189.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3167 | XP_016861678.1 | |
XM_017006190.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3166 | XP_016861679.1 | |
XM_017006191.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3131 | XP_016861680.1 | |
XM_017006192.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3128 | XP_016861681.1 | |
XM_017006193.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3127 | XP_016861682.1 | |
XM_017006194.1 | 9990 | Missense Mutation | GAC,GTC | D,V 3092 | XP_016861683.1 | |
XM_017006195.1 | 9990 | Missense Mutation | GAC,GTC | D,V 2919 | XP_016861684.1 |
HCLS1 - hematopoietic cell-specific Lyn substrate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |