Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTGGGCACCTGGATGTCCAAG[A/G]ACTATAGCAGGAGACAGGAGACAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603727 MIM: 614471 | |||||||||||||||||||||||
Literature Links: |
MIR6890 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MIR6890 - microRNA 6890 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QARS - glutaminyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272073.1 | 1836 | Missense Mutation | TCC,TTC | S,F 576 | NP_001259002.1 | |
NM_005051.2 | 1836 | Missense Mutation | TCC,TTC | S,F 587 | NP_005042.1 | |
XM_017006965.1 | 1836 | Missense Mutation | TCC,TTC | S,F 605 | XP_016862454.1 |
QRICH1 - glutamine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP19 - ubiquitin specific peptidase 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |