Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTCAGCTCCTGCCGCTCTTTC[A/G]TCCAGGCAGCCAGCACCTGCCTCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608560 | ||||||||||||||||||||
Literature Links: |
NT5DC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NT5DC2 - 5'-nucleotidase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134231.1 | 1511 | Missense Mutation | NP_001127703.1 | |||
NM_022908.2 | 1511 | Missense Mutation | NP_075059.1 | |||
XM_005265406.2 | 1511 | Missense Mutation | XP_005265463.1 | |||
XM_006713303.3 | 1511 | Missense Mutation | XP_006713366.1 | |||
XM_006713305.2 | 1511 | Missense Mutation | XP_006713368.1 |
STAB1 - stabilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015136.2 | 1511 | Intron | NP_055951.2 | |||
XM_005264973.3 | 1511 | Intron | XP_005265030.1 | |||
XM_005264974.2 | 1511 | Intron | XP_005265031.1 | |||
XM_006713065.1 | 1511 | Intron | XP_006713128.1 | |||
XM_017005998.1 | 1511 | Intron | XP_016861487.1 | |||
XM_017005999.1 | 1511 | Intron | XP_016861488.1 | |||
XM_017006000.1 | 1511 | Intron | XP_016861489.1 | |||
XM_017006001.1 | 1511 | Intron | XP_016861490.1 | |||
XM_017006002.1 | 1511 | Intron | XP_016861491.1 | |||
XM_017006003.1 | 1511 | Intron | XP_016861492.1 | |||
XM_017006004.1 | 1511 | Intron | XP_016861493.1 |