Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATCACCAGCACGGCCACAAAC[A/G]AGCAGCTGTTCACATAGGTGACTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606379 MIM: 611318 | |||||||||||||||||||||||
Literature Links: |
GPR87 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GPR87 - G protein-coupled receptor 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023915.3 | 1002 | Missense Mutation | TCG,TTG | S,L 214 | NP_076404.3 |
MED12L - mediator complex subunit 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053002.5 | 1002 | Intron | NP_443728.3 | |||
XM_006713487.3 | 1002 | Intron | XP_006713550.1 | |||
XM_011512390.2 | 1002 | Intron | XP_011510692.1 | |||
XM_011512394.2 | 1002 | Intron | XP_011510696.1 | |||
XM_011512399.2 | 1002 | Intron | XP_011510701.1 | |||
XM_017005676.1 | 1002 | Intron | XP_016861165.1 | |||
XM_017005677.1 | 1002 | Intron | XP_016861166.1 | |||
XM_017005678.1 | 1002 | Intron | XP_016861167.1 | |||
XM_017005679.1 | 1002 | Intron | XP_016861168.1 | |||
XM_017005680.1 | 1002 | Intron | XP_016861169.1 | |||
XM_017005681.1 | 1002 | Intron | XP_016861170.1 |