Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTAAACTGGTGTTTTAAAAGTA[A/G]GCTTTGGCTGGGTGCGGTGGCTCAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611295 | |||||||||||||||||||||||
Literature Links: |
KLHL24 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KLHL24 - kelch like family member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017644.3 | Intron | NP_060114.2 | ||||
XM_005247552.2 | Intron | XP_005247609.1 | ||||
XM_005247553.1 | Intron | XP_005247610.1 | ||||
XM_005247554.2 | Intron | XP_005247611.1 | ||||
XM_005247555.1 | Intron | XP_005247612.1 | ||||
XM_005247556.1 | Intron | XP_005247613.1 | ||||
XM_011512938.2 | Intron | XP_011511240.1 | ||||
XM_017006650.1 | Intron | XP_016862139.1 | ||||
XM_017006651.1 | Intron | XP_016862140.1 | ||||
XM_017006652.1 | Intron | XP_016862141.1 | ||||
XM_017006653.1 | Intron | XP_016862142.1 | ||||
XM_017006654.1 | Intron | XP_016862143.1 | ||||
XM_017006655.1 | Intron | XP_016862144.1 | ||||
XM_017006656.1 | Intron | XP_016862145.1 | ||||
XM_017006657.1 | Intron | XP_016862146.1 | ||||
XM_017006658.1 | Intron | XP_016862147.1 | ||||
XM_017006659.1 | Intron | XP_016862148.1 | ||||
XM_017006660.1 | Intron | XP_016862149.1 | ||||
XM_017006661.1 | Intron | XP_016862150.1 | ||||
XM_017006662.1 | Intron | XP_016862151.1 | ||||
XM_017006663.1 | Intron | XP_016862152.1 |