Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCTTTGAGCCTGGCATCCTGCT[A/G]TAAGTCTGGGTATTGCATGCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609679 | ||||||||||||||||||||
Literature Links: |
SLITRK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLITRK3 - SLIT and NTRK like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318810.1 | 3220 | Nonsense Mutation | CAG,TAG | Q,* 926 | NP_001305739.1 | |
NM_001318811.1 | 3220 | Nonsense Mutation | CAG,TAG | Q,* 926 | NP_001305740.1 | |
NM_014926.3 | 3220 | Nonsense Mutation | CAG,TAG | Q,* 926 | NP_055741.2 |