Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATCATTAGGAATGGATCTCTCA[C/T]TGTCTGTCCATGATTGGAGGATTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609916 MIM: 615166 | ||||||||||||||||||||
Literature Links: |
AZI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AZI2 - 5-azacytidine induced 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134432.1 | 1037 | Intron | NP_001127904.1 | |||
NM_001134433.1 | 1037 | Intron | NP_001127905.1 | |||
NM_001271650.1 | 1037 | Missense Mutation | AAT,AGT | N,S 210 | NP_001258579.1 | |
NM_022461.4 | 1037 | Missense Mutation | AAT,AGT | N,S 322 | NP_071906.1 | |
XM_005265392.2 | 1037 | Missense Mutation | AAT,AGT | N,S 322 | XP_005265449.1 | |
XM_011534009.2 | 1037 | Missense Mutation | AAT,AGT | N,S 325 | XP_011532311.1 | |
XM_011534010.2 | 1037 | Intron | XP_011532312.1 | |||
XM_011534011.2 | 1037 | Intron | XP_011532313.1 |
CMC1 - C-X9-C motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |