Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTTCATGGGTGTTTGTCCTGC[A/G]TTTTCTCCTATAACAAAATGAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607392 | ||||||||||||||||||||
Literature Links: |
WWTR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WWTR1 - WW domain containing transcription regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168278.1 | 1230 | Silent Mutation | AAC,AAT | N,N 342 | NP_001161750.1 | |
NM_001168280.1 | 1230 | Silent Mutation | AAC,AAT | N,N 342 | NP_001161752.1 | |
NM_015472.4 | 1230 | Silent Mutation | AAC,AAT | N,N 342 | NP_056287.1 | |
XM_017006121.1 | 1230 | Silent Mutation | AAC,AAT | N,N 342 | XP_016861610.1 | |
XM_017006122.1 | 1230 | Silent Mutation | AAC,AAT | N,N 342 | XP_016861611.1 |