Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCAGGAACCTACCAGGTGCAG[C/T]TCATCACCCTCGATCCACTGGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180260 | ||||||||||||||||||||
Literature Links: |
LOC100507291 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507291 - uncharacterized LOC100507291 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBP1 - retinol binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130992.1 | 642 | Intron | NP_001124464.1 | |||
NM_001130993.1 | 642 | Intron | NP_001124465.1 | |||
NM_002899.3 | 642 | Silent Mutation | GAA,GAG | E,E 177 | NP_002890.2 |