Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAAGCCCCAGAATATAACCAA[C/T]CATTGACATTACTGCAGTGGAACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601215 | ||||||||||||||||||||
Literature Links: |
ATR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATR - ATR serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184.3 | 7296 | Missense Mutation | ATT,GTT | I,V 2467 | NP_001175.2 | |
XM_011512924.1 | 7296 | Missense Mutation | ATT,GTT | I,V 2469 | XP_011511226.1 | |
XM_011512925.1 | 7296 | Missense Mutation | ATT,GTT | I,V 2405 | XP_011511227.1 | |
XM_011512926.2 | 7296 | Intron | XP_011511228.1 | |||
XM_017006643.1 | 7296 | Missense Mutation | ATT,GTT | I,V 2403 | XP_016862132.1 |