Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGGTGCTGGCACTGGCCTCCA[C/T]GGTCACAGTAGCCCCTACTGCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 158372 | ||||||||||||||||||||
Literature Links: |
MUC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUC4 - mucin 4, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322468.1 | 22166 | Missense Mutation | NP_001309397.1 | |||
NM_004532.5 | 22166 | Missense Mutation | CAT,CGT | H,R 1095 | NP_004523.3 | |
NM_018406.6 | 22166 | Intron | NP_060876.5 | |||
NM_138297.4 | 22166 | Missense Mutation | CAT,CGT | H,R 1044 | NP_612154.2 |