Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGCAAGTGTGATGCATCCTCC[A/G]CCCAAAAAATGGAACCTAAATAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607002 | |||||||||||||||||||||||
Literature Links: |
PROK2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PROK2 - prokineticin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126128.1 | 392 | Missense Mutation | CGG,TGG | R,W 101 | NP_001119600.1 | |
NM_021935.3 | 392 | Missense Mutation | CGG,TGG | R,W 80 | NP_068754.1 | |
XM_017006974.1 | 392 | Missense Mutation | CGG,TGG | R,W 81 | XP_016862463.1 |