Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATAAGGTTGGCATTTGGATTAT[C/T]AGTCCCTGGTCCTGTTGGACTCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608628 | ||||||||||||||||||||
Literature Links: |
TBL1XR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBL1XR1 - transducin (beta)-like 1 X-linked receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321193.1 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | NP_001308122.1 | |
NM_001321194.1 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | NP_001308123.1 | |
NM_001321195.1 | 1396 | Missense Mutation | AAT,GAT | N,D 320 | NP_001308124.1 | |
NM_024665.5 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | NP_078941.2 | |
XM_005247775.2 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_005247832.1 | |
XM_006713746.1 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_006713809.1 | |
XM_011513141.1 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_011511443.1 | |
XM_011513142.2 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_011511444.1 | |
XM_011513143.2 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_011511445.1 | |
XM_017007185.1 | 1396 | Missense Mutation | AAT,GAT | N,D 407 | XP_016862674.1 |