Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAACAGGACTTTTCTCTCTG[C/T]CACACCCTCTCCTAGGAACACCCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615507 | |||||||||||||||||||||||
Literature Links: |
NISCH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NISCH - nischarin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276293.1 | Intron | NP_001263222.1 | ||||
NM_001276294.1 | Intron | NP_001263223.1 | ||||
NM_007184.3 | Intron | NP_009115.2 | ||||
XM_005264839.2 | Intron | XP_005264896.1 | ||||
XM_006712955.2 | Intron | XP_006713018.1 | ||||
XM_017005643.1 | Intron | XP_016861132.1 |