Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCACTGCTTGCCATGCCTGAG[A/G]AGACCCAAGAAGGTAGGAGAGGGTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602725 MIM: 601281 | |||||||||||||||||||||||
Literature Links: |
IFRD2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IFRD2 - interferon related developmental regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LSMEM2 - leucine rich single-pass membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304385.1 | 133 | Missense Mutation | AAG,GAG | K,E 16 | NP_001291314.1 | |
NM_153215.2 | 133 | Missense Mutation | AAG,GAG | K,E 16 | NP_694947.1 | |
XM_006712979.3 | 133 | Intron | XP_006713042.1 | |||
XM_006712980.3 | 133 | Intron | XP_006713043.1 | |||
XM_011533370.2 | 133 | Intron | XP_011531672.1 |
MIR6872 - microRNA 6872 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA3B - semaphorin 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |