Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAGCTAGTAACTTCAAATTTCA[C/T]AAAACATTATCTTTCATTGAAGGGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610819 | |||||||||||||||||||||||
Literature Links: |
SLC25A38 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC25A38 - solute carrier family 25 member 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017875.2 | Intron | NP_060345.2 | ||||
XM_006713214.1 | Intron | XP_006713277.1 | ||||
XM_011533869.2 | Intron | XP_011532171.1 | ||||
XM_017006710.1 | Intron | XP_016862199.1 |