Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTCCTGAGCGTGGTTGGTGGGG[A/G]CTCTAAGGCCATTCCTCCAGGCCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601460 | |||||||||||||||||||||||
Literature Links: |
C3orf36 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf36 - chromosome 3 open reading frame 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025041.2 | 1151 | Missense Mutation | CCC,TCC | P,S 48 | NP_079317.2 |
SLCO2A1 - solute carrier organic anion transporter family member 2A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |