Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATCCCATCAACGTTAAGATGGA[C/G]AGACCAGCCCAAATCTTTAGTTGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607474 | |||||||||||||||||||||||
Literature Links: |
HGD PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HGD - homogentisate 1,2-dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABL3 - RAB, member of RAS oncogene family like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173825.3 | 3380 | UTR 3 | NP_776186.2 | |||
XM_006713595.2 | 3380 | UTR 3 | XP_006713658.1 | |||
XM_017006227.1 | 3380 | UTR 3 | XP_016861716.1 | |||
XM_017006228.1 | 3380 | UTR 3 | XP_016861717.1 |