Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTCTCTCTCAAACAAGCCTTT[A/C]TTATTCTGAAAGATGTCTTTTATCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609980 | |||||||||||||||||||||||
Literature Links: |
VGLL3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
VGLL3 - vestigial like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320493.1 | 6085 | Intron | NP_001307422.1 | |||
NM_001320494.1 | 6085 | UTR 3 | NP_001307423.1 | |||
NM_016206.3 | 6085 | UTR 3 | NP_057290.2 | |||
XM_006713138.3 | 6085 | UTR 3 | XP_006713201.1 |