Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCAGCGTCCTGACTAGATCCGG[A/G]GATTGCATTAGACACAGCAGTACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TSC22D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSC22D2 - TSC22 domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303264.1 | 1613 | Silent Mutation | GGA,GGG | G,G 199 | NP_001290193.1 | |
NM_014779.3 | 1613 | Silent Mutation | GGA,GGG | G,G 199 | NP_055594.1 | |
XM_011513337.2 | 1613 | Silent Mutation | GGA,GGG | G,G 199 | XP_011511639.1 |