Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAAAGAGGAGAAGTTGCAGATG[C/T]AAATGTATTAAAGTGAAGTGTGTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607640 MIM: 611965 | |||||||||||||||||||||||
Literature Links: |
ATXN7 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATXN7 - ataxin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000333.3 | Intron | NP_000324.1 | ||||
NM_001128149.2 | Intron | NP_001121621.2 | ||||
NM_001177387.1 | Intron | NP_001170858.1 |
THOC7 - THO complex 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285387.2 | Intron | NP_001272316.1 | ||||
NM_001285404.1 | Intron | NP_001272333.1 | ||||
NM_025075.3 | Intron | NP_079351.2 | ||||
XM_006713339.3 | Intron | XP_006713402.1 |
THOC7-AS1 - THOC7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |