Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGGTGGGCCAGATGTCATCAA[C/G]TGGGAAAGGAGAGTAATTCCCGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604544 | ||||||||||||||||||||
Literature Links: |
LARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LARS2 - leucyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015340.3 | 266 | Missense Mutation | AAC,AAG | N,K 27 | NP_056155.1 | |
XM_005265006.1 | 266 | Missense Mutation | AAC,AAG | N,K 27 | XP_005265063.1 | |
XM_011533554.2 | 266 | Missense Mutation | AAC,AAG | N,K 27 | XP_011531856.1 | |
XM_017006042.1 | 266 | Missense Mutation | AAC,AAG | N,K 27 | XP_016861531.1 |