Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGATTTTCCAAATAGTGTGGCA[C/T]GTTGTAAATAAAGGCTTCATTATGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607156 | |||||||||||||||||||||||
Literature Links: |
ST3GAL6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ST3GAL6 - ST3 beta-galactoside alpha-2,3-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271142.1 | Intron | NP_001258071.1 | ||||
NM_001271145.1 | Intron | NP_001258074.1 | ||||
NM_001271146.1 | Intron | NP_001258075.1 | ||||
NM_001271147.1 | Intron | NP_001258076.1 | ||||
NM_001271148.1 | Intron | NP_001258077.1 | ||||
NM_001323352.1 | Intron | NP_001310281.1 | ||||
NM_001323353.1 | Intron | NP_001310282.1 | ||||
NM_001323358.1 | Intron | NP_001310287.1 | ||||
NM_001323359.1 | Intron | NP_001310288.1 | ||||
NM_001323360.1 | Intron | NP_001310289.1 | ||||
NM_001323362.1 | Intron | NP_001310291.1 | ||||
NM_001323363.1 | Intron | NP_001310292.1 | ||||
NM_001323364.1 | Intron | NP_001310293.1 | ||||
NM_001323365.1 | Intron | NP_001310294.1 | ||||
NM_001323366.1 | Intron | NP_001310295.1 | ||||
NM_001323367.1 | Intron | NP_001310296.1 | ||||
NM_001323368.1 | Intron | NP_001310297.1 | ||||
NM_006100.3 | Intron | NP_006091.1 |