Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCAGGACCTATGACAGTGCCTG[G/T]CACTGAGTTGGTGCTCCATGAATAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613726 | |||||||||||||||||||||||
Literature Links: |
ANO10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ANO10 - anoctamin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204831.1 | 2026 | Missense Mutation | NP_001191760.1 | |||
NM_001204832.1 | 2026 | Intron | NP_001191761.1 | |||
NM_001204833.1 | 2026 | Intron | NP_001191762.1 | |||
NM_001204834.1 | 2026 | Intron | NP_001191763.1 | |||
NM_018075.3 | 2026 | Intron | NP_060545.3 | |||
XM_011533885.2 | 2026 | Intron | XP_011532187.2 | |||
XM_011533887.2 | 2026 | Intron | XP_011532189.1 | |||
XM_011533889.2 | 2026 | Intron | XP_011532191.1 | |||
XM_011533890.2 | 2026 | Intron | XP_011532192.1 | |||
XM_017006716.1 | 2026 | Intron | XP_016862205.1 | |||
XM_017006717.1 | 2026 | Intron | XP_016862206.1 | |||
XM_017006718.1 | 2026 | Intron | XP_016862207.1 | |||
XM_017006719.1 | 2026 | Intron | XP_016862208.1 | |||
XM_017006720.1 | 2026 | Intron | XP_016862209.1 | |||
XM_017006721.1 | 2026 | Intron | XP_016862210.1 | |||
XM_017006722.1 | 2026 | Intron | XP_016862211.1 |