Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGATTTAACAGCCTTAATGGT[C/T]GATGAAATAAGAACTGTAAGGAAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TMEM39A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM39A - transmembrane protein 39A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018266.2 | 1512 | Missense Mutation | CAA,CGA | Q,R 416 | NP_060736.1 | |
XM_005247578.1 | 1512 | Missense Mutation | AAC,GAC | N,D 376 | XP_005247635.1 | |
XM_006713687.1 | 1512 | Missense Mutation | CAA,CGA | Q,R 416 | XP_006713750.1 | |
XM_011512958.2 | 1512 | Intron | XP_011511260.1 | |||
XM_017006788.1 | 1512 | Missense Mutation | CAA,CGA | Q,R 388 | XP_016862277.1 |