Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACTCCTTGTCAGACAGTGACAG[A/C]ATCATAGCATATGCCTGGGGTAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 165215 | |||||||||||||||||||||||
Literature Links: |
MECOM PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MECOM - MDS1 and EVI1 complex locus | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105077.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1077 | NP_001098547.3 | |
NM_001105078.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1012 | NP_001098548.2 | |
NM_001163999.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1004 | NP_001157471.1 | |
NM_001164000.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1003 | NP_001157472.1 | |
NM_001205194.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1012 | NP_001192123.1 | |
NM_004991.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1200 | NP_004982.2 | |
NM_005241.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1012 | NP_005232.2 | |
XM_005247213.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1201 | XP_005247270.1 | |
XM_005247214.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1192 | XP_005247271.1 | |
XM_005247215.3 | 3301 | Missense Mutation | ATG,ATT | M,I 1191 | XP_005247272.1 | |
XM_005247219.2 | 3301 | Missense Mutation | ATG,ATT | M,I 1013 | XP_005247276.1 | |
XM_005247220.2 | 3301 | Missense Mutation | ATG,ATT | M,I 1013 | XP_005247277.1 | |
XM_005247221.2 | 3301 | Missense Mutation | ATG,ATT | M,I 1013 | XP_005247278.1 | |
XM_005247223.2 | 3301 | Missense Mutation | ATG,ATT | M,I 1012 | XP_005247280.1 | |
XM_005247224.3 | 3301 | Missense Mutation | ATG,ATT | M,I 877 | XP_005247281.1 | |
XM_005247225.3 | 3301 | Missense Mutation | ATG,ATT | M,I 876 | XP_005247282.1 | |
XM_005247226.3 | 3301 | Missense Mutation | ATG,ATT | M,I 867 | XP_005247283.1 | |
XM_011512546.2 | 3301 | Missense Mutation | ATG,ATT | M,I 1085 | XP_011510848.1 | |
XM_017005874.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1077 | XP_016861363.1 | |
XM_017005875.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1003 | XP_016861364.1 | |
XM_017005876.1 | 3301 | Missense Mutation | ATG,ATT | M,I 1004 | XP_016861365.1 | |
XM_017005877.1 | 3301 | Missense Mutation | ATG,ATT | M,I 868 | XP_016861366.1 | |
XM_017005878.1 | 3301 | Missense Mutation | ATG,ATT | M,I 679 | XP_016861367.1 |