Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACAGTACATGAGCGAATCCACA[C/T]GGGAGAGAAACCCTATAAGTGTAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF660 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF660 - zinc finger protein 660 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173658.2 | 551 | Missense Mutation | ACG,ATG | T,M 73 | NP_775929.2 | |
XM_005265090.4 | 551 | Missense Mutation | ACG,ATG | T,M 73 | XP_005265147.1 | |
XM_006713117.3 | 551 | Missense Mutation | ACG,ATG | T,M 73 | XP_006713180.1 | |
XM_011533622.2 | 551 | Missense Mutation | ACG,ATG | T,M 73 | XP_011531924.1 |