Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTCAGCCTCACAAGTTGGGGG[G/T]GGCCCAATTAAGGAACTTCATGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176910 MIM: 613698 | ||||||||||||||||||||
Literature Links: |
PRKAR2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKAR2A - protein kinase cAMP-dependent type II regulatory subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAR2A-AS1 - PRKAR2A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A20 - solute carrier family 25 member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000387.5 | 1091 | Missense Mutation | ACC,CCC | T,P 298 | NP_000378.1 |