Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCCTTTTCCTTAGGTGGGGCAC[G/T]AGCGTAAGGTCCAGCTGGTCACCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605615 MIM: 614584 | ||||||||||||||||||||
Literature Links: |
ARIH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARIH2 - ariadne RBR E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P4HTM - prolyl 4-hydroxylase, transmembrane | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177938.2 | 713 | Nonsense Mutation | GAG,TAG | E,* 122 | NP_808807.2 | |
NM_177939.2 | 713 | Nonsense Mutation | GAG,TAG | E,* 122 | NP_808808.1 |