Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCAAGGGGAACTGCCATGGGGC[C/T]TCGATCCCTGGGAACCTATGGGCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF620 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF620 - zinc finger protein 620 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256167.1 | 363 | Intron | NP_001243096.1 | |||
NM_001256168.1 | 363 | Intron | NP_001243097.1 | |||
NM_175888.3 | 363 | Missense Mutation | CTC,TTC | L,F 72 | NP_787084.1 | |
XM_005265011.3 | 363 | Missense Mutation | CTC,TTC | L,F 72 | XP_005265068.1 | |
XM_005265012.3 | 363 | Missense Mutation | CTC,TTC | L,F 72 | XP_005265069.1 | |
XM_017006064.1 | 363 | Missense Mutation | CTC,TTC | L,F 117 | XP_016861553.1 | |
XM_017006065.1 | 363 | Intron | XP_016861554.1 | |||
XM_017006066.1 | 363 | Missense Mutation | CTC,TTC | L,F 33 | XP_016861555.1 | |
XM_017006067.1 | 363 | Missense Mutation | CTC,TTC | L,F 33 | XP_016861556.1 | |
XM_017006068.1 | 363 | Intron | XP_016861557.1 | |||
XM_017006069.1 | 363 | Intron | XP_016861558.1 |