Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCGGTATCCAGGCCTGTTTTGA[A/G]CGCGCCTCGGAGTACCAGCTCAACG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 139330 MIM: 601124 | |||||||||||||||||||||||
Literature Links: |
GNAT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GNAT1 - G protein subunit alpha transducin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000172.3 | 527 | Silent Mutation | GAA,GAG | E,E 137 | NP_000163.2 | |
NM_144499.2 | 527 | Silent Mutation | GAA,GAG | E,E 137 | NP_653082.1 |
SEMA3F - semaphorin 3F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |