Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTTGCTGCCCAAATTTTTGAAC[A/T]TTATCTGCCAAATTGTTAATACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608922 | ||||||||||||||||||||
Literature Links: |
ARL13B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL13B - ADP ribosylation factor like GTPase 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174150.1 | 458 | Intron | NP_001167621.1 | |||
NM_001174151.1 | 458 | Intron | NP_001167622.1 | |||
NM_001321328.1 | 458 | Intron | NP_001308257.1 | |||
NM_144996.3 | 458 | Intron | NP_659433.2 | |||
NM_182896.2 | 458 | Intron | NP_878899.1 | |||
XM_006713532.3 | 458 | Intron | XP_006713595.1 | |||
XM_011512532.2 | 458 | Intron | XP_011510834.1 | |||
XM_011512533.2 | 458 | Intron | XP_011510835.1 | |||
XM_011512534.2 | 458 | Intron | XP_011510836.1 | |||
XM_011512535.2 | 458 | Intron | XP_011510837.1 | |||
XM_017005853.1 | 458 | Intron | XP_016861342.1 |
STX19 - syntaxin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001850.2 | 458 | Silent Mutation | AAA,AAT | K,N 67 | NP_001001850.1 | |
XM_011512842.2 | 458 | Missense Mutation | AAA,AAT | K,N 67 | XP_011511144.1 |