Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGAGAATTCTCTAAGCATCATG[C/T]GAAAAGCTGTCAACCTGGGCAATAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614724 MIM: 605739 | |||||||||||||||||||||||
Literature Links: |
CEP63 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KY - kyphoscoliosis peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178554.4 | 7466 | UTR 3 | NP_848649.3 | |||
XM_005247418.3 | 7466 | UTR 3 | XP_005247475.1 | |||
XM_006713612.3 | 7466 | UTR 3 | XP_006713675.1 | |||
XM_017006287.1 | 7466 | UTR 3 | XP_016861776.1 | |||
XM_017006288.1 | 7466 | UTR 3 | XP_016861777.1 | |||
XM_017006289.1 | 7466 | UTR 3 | XP_016861778.1 | |||
XM_017006290.1 | 7466 | UTR 3 | XP_016861779.1 |
LOC101927568 - uncharacterized LOC101927568 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |