Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATATAGCCAAGGGTTAATGACCT[A/G]TATGATTCCAGCAGCTGACTGTATG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
PSMD6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PSMD6 - proteasome 26S subunit, non-ATPase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271779.1 | 1120 | Silent Mutation | TAC,TAT | Y,Y 364 | NP_001258708.1 | |
NM_001271780.1 | 1120 | Silent Mutation | TAC,TAT | Y,Y 273 | NP_001258709.1 | |
NM_001271781.1 | 1120 | Silent Mutation | TAC,TAT | Y,Y 272 | NP_001258710.1 | |
NM_014814.2 | 1120 | Silent Mutation | TAC,TAT | Y,Y 311 | NP_055629.1 | |
XM_005265619.1 | 1120 | Silent Mutation | TAC,TAT | Y,Y 262 | XP_005265676.1 | |
XM_011534288.1 | 1120 | Intron | XP_011532590.1 | |||
XM_017007569.1 | 1120 | Silent Mutation | TAC,TAT | Y,Y 266 | XP_016863058.1 |
PSMD6-AS2 - PSMD6 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |