Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTACACAGTCTCTGTAGCT[C/T]CTAGGAATTTCATTTCTACAGCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 138160 | |||||||||||||||||||||||
Literature Links: |
SLC2A2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC2A2 - solute carrier family 2 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000340.1 | 1865 | Missense Mutation | GAA,GGA | E,G 519 | NP_000331.1 | |
NM_001278658.1 | 1865 | Missense Mutation | GAA,GGA | E,G 400 | NP_001265587.1 | |
NM_001278659.1 | 1865 | Missense Mutation | GAA,GGA | E,G 346 | NP_001265588.1 | |
XM_011513087.2 | 1865 | Missense Mutation | GAA,GGA | E,G 504 | XP_011511389.1 | |
XM_011513089.2 | 1865 | Missense Mutation | GAA,GGA | E,G 346 | XP_011511391.1 |