Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTACTAGAGTACCGAGAAGCCTG[C/T]GCAGCCAAAAAGTTCTCCTCAGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616709 | ||||||||||||||||||||
Literature Links: |
A4GNT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
A4GNT - alpha-1,4-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016161.2 | 754 | Silent Mutation | GCA,GCG | A,A 184 | NP_057245.1 | |
XM_017006543.1 | 754 | Silent Mutation | GCA,GCG | A,A 184 | XP_016862032.1 | |
XM_017006544.1 | 754 | Silent Mutation | GCA,GCG | A,A 184 | XP_016862033.1 |
DZIP1L - DAZ interacting zinc finger protein 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |