Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTAAAATAAGCAGGTGGATCTA[G/T]TTCATGTTTTTGATCAAAAACTATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 116806 MIM: 617010 | |||||||||||||||||||||||
Literature Links: |
CTNNB1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CTNNB1 - catenin beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098209.1 | 2906 | UTR 3 | NP_001091679.1 | |||
NM_001098210.1 | 2906 | UTR 3 | NP_001091680.1 | |||
NM_001904.3 | 2906 | UTR 3 | NP_001895.1 | |||
XM_005264886.2 | 2906 | UTR 3 | XP_005264943.1 | |||
XM_006712983.1 | 2906 | UTR 3 | XP_006713046.1 | |||
XM_006712984.1 | 2906 | UTR 3 | XP_006713047.1 | |||
XM_006712985.1 | 2906 | UTR 3 | XP_006713048.1 | |||
XM_017005738.1 | 2906 | UTR 3 | XP_016861227.1 |
ULK4 - unc-51 like kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |