Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATCACAGGGTTGAGGGCGCTAT[C/T]CACGTAGCCCAGCCATGTCGTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126451 | ||||||||||||||||||||
Literature Links: |
DRD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRD3 - dopamine receptor D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000796.5 | 1546 | Missense Mutation | AAT,GAT | N,D 375 | NP_000787.2 | |
NM_001282563.2 | 1546 | Missense Mutation | AAT,GAT | N,D 375 | NP_001269492.1 | |
NM_001290809.1 | 1546 | Missense Mutation | AAT,GAT | N,D 375 | NP_001277738.1 | |
NM_033663.5 | 1546 | Missense Mutation | AAT,GAT | N,D 342 | NP_387512.3 | |
XM_017005829.1 | 1546 | Missense Mutation | AAT,GAT | N,D 375 | XP_016861318.1 |