Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCATCTCAATGTCAGTGATTAC[A/G]TCGTCCGGATCAAAGGAAAGCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609107 MIM: 601306 | ||||||||||||||||||||
Literature Links: |
FBXO40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO40 - F-box protein 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCLS1 - hematopoietic cell-specific Lyn substrate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292041.1 | 1377 | Silent Mutation | GAC,GAT | D,D 416 | NP_001278970.1 | |
NM_005335.5 | 1377 | Silent Mutation | GAC,GAT | D,D 453 | NP_005326.2 |