Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTCTGAGGACATCATGCTCACA[C/T]GTCAGAATGTAGGTCTTTGGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613234 | ||||||||||||||||||||
Literature Links: |
NCEH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCEH1 - neutral cholesterol ester hydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146276.1 | 1266 | Silent Mutation | ACA,ACG | T,T 384 | NP_001139748.1 | |
NM_001146277.1 | 1266 | Silent Mutation | ACA,ACG | T,T 211 | NP_001139749.1 | |
NM_001146278.1 | 1266 | Silent Mutation | ACA,ACG | T,T 211 | NP_001139750.1 | |
NM_020792.4 | 1266 | Silent Mutation | ACA,ACG | T,T 376 | NP_065843.3 |