Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCGACTCGCATTTCGCATTAAG[A/G]AACTGGAAAACCTAGAGTGACAAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604180 | |||||||||||||||||||||||
Literature Links: |
LOC105374013 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC105374013 - endogenous retrovirus group K member 5 Gag polyprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011513341.2 | Intron | XP_011511643.1 |
RPL24 - ribosomal protein L24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000986.3 | Intron | NP_000977.1 |
ZBTB11 - zinc finger and BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB11-AS1 - ZBTB11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |