Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCTTCATGTCTCACTTTAGG[C/T]TGTCGATGGCGTGTTCATCCGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147270 MIM: 601959 | ||||||||||||||||||||
Literature Links: |
ITIH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITIH1 - inter-alpha-trypsin inhibitor heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166434.2 | 170 | Intron | NP_001159906.1 | |||
NM_001166435.2 | 170 | Intron | NP_001159907.1 | |||
NM_001166436.2 | 170 | Intron | NP_001159908.1 | |||
NM_002215.3 | 170 | Missense Mutation | GCT,GTT | A,V 47 | NP_002206.2 |
NEK4 - NIMA related kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |