Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAAGGAGGAGGAATATGGAATC[C/T]AAGGGGGCCAGTTCCTGCCGTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601662 | ||||||||||||||||||||
Literature Links: |
ALCAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALCAM - activated leukocyte cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243280.1 | 705 | Silent Mutation | TCC,TCT | S,S 3 | NP_001230209.1 | |
NM_001243281.1 | 705 | Silent Mutation | TCC,TCT | S,S 3 | NP_001230210.1 | |
NM_001243283.1 | 705 | Silent Mutation | TCC,TCT | S,S 3 | NP_001230212.1 | |
NM_001627.3 | 705 | Silent Mutation | TCC,TCT | S,S 3 | NP_001618.2 |