Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTAATTAGGGATTTGCAATTA[G/T]CTAGGCAAGGCTAGGATAAAGTTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611909 | |||||||||||||||||||||||
Literature Links: |
FNDC3B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FNDC3B - fibronectin type III domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135095.1 | Intron | NP_001128567.1 | ||||
NM_022763.3 | Intron | NP_073600.3 | ||||
XM_017007062.1 | Intron | XP_016862551.1 | ||||
XM_017007063.1 | Intron | XP_016862552.1 | ||||
XM_017007064.1 | Intron | XP_016862553.1 |